Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Methylbiphenyl-4-carboxaldehyde, 96%

Catalog No. AAH3424903
Click to view available options
Quantity:
250 mg
1 g

CAS: 36393-42-7 | C14H12O | 196.249 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36393-42-7
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.249
MDL Number MFCD02258938
InChI Key BCINBWXQYBLSKO-UHFFFAOYSA-N
Synonym 4-4-methylphenyl benzaldehyde, 4'-methyl-1,1'-biphenyl-4-carbaldehyde, 4'-methylbiphenyl-4-carbaldehyde, 4'-methyl-4-formylbiphenyl, 1,1'-biphenyl-4-carboxaldehyde, 4'-methyl, 4'-methyl-biphenyl-4-carbaldehyde, 4'-methylbiphenyl-4-carboxaldehyde, 4'-methyl-biphenyl-4-carboxaldehyde, 4'-methyl 1,1'-biphenyl-4-carbaldehyde, pubchem10296
PubChem CID 1392762
IUPAC Name 4-(4-methylphenyl)benzaldehyde
SMILES CC1=CC=C(C=C1)C2=CC=C(C=C2)C=O

Specifications

Melting Point 108°C to 113°C
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 196.25
Percent Purity 96%
Chemical Name or Material 4'-Methylbiphenyl-4-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.