Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylcyclohexanecarboxylic Acid (cis- and trans- mixture) 98.0+%, TCI America™
SDP

Catalog No. M162125G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 4331-54-8
Molecular Formula C8H14O2
Molecular Weight (g/mol) 142.20
MDL Number MFCD00074943,MFCD00074909,MFCD20617670
InChI Key QTDXSEZXAPHVBI-UHFFFAOYSA-N
PubChem CID 20330
IUPAC Name 4-methylcyclohexane-1-carboxylic acid
SMILES CC1CCC(CC1)C(O)=O

Specifications

Color Colorless
Boiling Point 140°C
Quantity 25 g
Formula Weight 142.20
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-Methylcyclohexanecarboxylic Acid (cis- and trans- mixture)
EINECSNumber (3)-3311
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.