4-Methylcyclohexanol, cis + trans, 98%

Manufacturer: Alfa Aesar A16734AD

Quantity 50mL

 View more versions of this product

Catalog No. AAA16734AD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 589-91-3
    Molecular Formula C7H14O
    Formula Weight 114.19
    MDL Number MFCD00001449
    Beilstein 1445998
    Chemical Name or Material 4-Methylcyclohexanol cis + trans, cis + trans

    Specifications

    Formula Weight 114.19
    Quantity 50mL
    TSCA TSCA
    MDL Number MFCD00001449
    Beilstein 1445998
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H227
    Combustible liquid.
    Health Hazard 3 GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction:
    Molecular Formula C7H14O
    CAS 589-91-3
    Chemical Name or Material 4-Methylcyclohexanol cis + trans, cis + trans
    Assay 98%

    Description

    Applications
    2-Methylcyclohexanol was used to study the effect of organic solvents on epoxide hydrolase.

    Solubility
    Soluble in water (15 mg/ml at 20°C), ether, alcohols, and many organic solvents.

    Notes
    Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acid anhydrides, acid chlorides.

    GHS H Statement
    H227
    Combustible liquid.

    GHS P Statement
    P210-P280-P370+P378a-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    In case of fire: Use for extinction:

    Warning

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit