Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methyldiphenylamine, 98%

Catalog No. p-7059385
Click to view available options
Quantity:
10 g
50 g

CAS: 620-84-8 | C13H13N | 183.25 g/mol

4-Methyldiphenylamine is used as a syntheses material intermediates as well as OLED materials. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-84-8
Molecular Formula C13H13N
Molecular Weight (g/mol) 183.25
MDL Number MFCD00092921
InChI Key AGHYMXKKEXDUTA-UHFFFAOYSA-N
Synonym 4-methyldiphenylamine, benzenamine, 4-methyl-n-phenyl, n-phenyl-p-toluidine, 4-methyl-n-phenyl-aniline, 4-methyl-n-phenylbenzamine, 4-methylphenyl phenylamine, phenyl-p-tolylamine, 4-methyldiphenylamin, phenyl-p-tolyl-amine, n-pnenyl-4-methylaniline
PubChem CID 12109
IUPAC Name 4-methyl-N-phenylaniline
SMILES CC1=CC=C(NC2=CC=CC=C2)C=C1

Specifications

Melting Point 85°C to 86°C
Color Brown
Boiling Point 334°C
Flash Point 155°C (311°F)
Quantity 10 g
Solubility Information Insoluble in water. Soluble in benzene,toluene,methanol,ethanol
Formula Weight 183.25
Percent Purity 98%
Physical Form Crystalline
Chemical Name or Material 4-Methyldiphenylamine
Hazard Category H302+H312-H315-H319
Hazard Statement GHS H Statement
H302-H312-H315-H319
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 210-655-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.