Learn More
4-Methylimidazole, 98%
Description
4-Methylimidazole is used for the curing agent of epoxy resin, the main raw material of medicine cimetidine. It can also be used for synthesis of antibacterial agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Methylimidazole is used for the curing agent of epoxy resin, the main raw material of medicine cimetidine. It can also be used for synthesis of antibacterial agent.
Solubility
Soluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.
Specifications
Specifications
| Melting Point | 49°C to 57°C |
| Density | 1.02 |
| Boiling Point | 263°C |
| Flash Point | 157°C (314°F) |
| Quantity | 50 g |
| UN Number | UN3263 |
| Beilstein | 1453 |
| Solubility Information | Soluble in water. |
| Formula Weight | 82.11 |
| Percent Purity | 98% |
| Show More |
Safety and Handling
| Hazard Category | H302-H311-H314-H335-H350 |
| Hazard Statement | GHS H Statement H314-H318-H302 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. |
| Precautionary Statement | P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Hazard Class: 6.1; Packaging Group: III |
| EINECSNumber | 212-497-3 |
| RTECSNumber | NI7350000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.