Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylindole, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
16096-32-5
C9H9N
131.18
MFCD00005668
PZOUSPYUWWUPPK-UHFFFAOYSA-N
4-methylindole,4-methyindole,1h-indole, 4-methyl,indole, 4-methyl,4-methyl indole,4-methyl-indole,pubchem7233,acmc-1bpu3,ksc179o0l
85282
4-methyl-1H-indole
CC1=C2C=CNC2=CC=C1
16096-32-5
C9H9N
131.18
MFCD00005668
PZOUSPYUWWUPPK-UHFFFAOYSA-N
4-methylindole,4-methyindole,1h-indole, 4-methyl,indole, 4-methyl,4-methyl indole,4-methyl-indole,pubchem7233,acmc-1bpu3,ksc179o0l
85282
4-methyl-1H-indole
CC1=C2C=CNC2=CC=C1
Melting Point 5°C
Boiling Point 267°C
Flash Point >112°C
Specific Gravity 1.06
Refractive Index 1.6060 to 1.608
Density 1.0600g/mL
Infrared Spectrum Authentic
Quantity 1g
Packaging Glass bottle
Color Yellow to Brown
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 20,II,204
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,ethyl ether and acetone
Formula Weight 131.18
Physical Form Liquid After Melting
Percent Purity ≥98.5%
Chemical Name or Material 4-Methylindole, 99%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 240-262-5

Recommended Storage : This product darkens on exposure to light