Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrakis(hydroxymethyl)phosphonium chloride, approx. 75-85% solution in water

Catalog No. AC389448000
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
Glass bottle

CAS: 124-64-1 | C4H12ClO4P | 190.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tetrakis(hydroxymethyl)phosphonium chloride
CAS 124-64-1, 7732-18-5
Color Green-Yellow or Pink
Density 1.3400g/mL
Boiling Point 115.0°C
Flash Point 96°C
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C4H12ClO4P
Quantity 800 mL
MDL Number MFCD00031687
Specific Gravity 1.34
Synonym tetrakis hydroxymethyl phosphonium chloride, thpc, pyroset tkc, retardol c, tetramethylolphosphonium chloride, tetrakis hydroxymethyl phosphanium chloride, unii-58wb2xcf8i, proban cc, tetrakis hydroxymethyl phosphochloride, ccris 317
Solubility Information Solubility in water: soluble.
InChI Key AKXUUJCMWZFYMV-UHFFFAOYSA-M
SMILES [Cl-].OC[P+](CO)(CO)CO
IUPAC Name tetrakis(hydroxymethyl)phosphanium;chloride
Molecular Weight (g/mol) 190.56
PubChem CID 31298
Formula Weight 190.56
Percent Purity 80.0 to 85.0%
Physical Form Solution
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.