Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylphenoxyacetic Acid 98.0+%, TCI America™
SDP

Catalog No. M272925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M272925G 25 g
M27295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M272925G Supplier TCI America Supplier No. M272925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 940-64-7
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00014365
InChI Key SFTDDFBJWUWKMN-UHFFFAOYSA-N
Synonym 4-methylphenoxy acetic acid, 4-methylphenoxyacetic acid, 2-4-methylphenoxy acetic acid, p-methylphenoxyacetic acid, unii-wwy23322ik, acetic acid, 4-methylphenoxy, 2-p-tolyloxy acetic acid, p-toloxyacetic acid, acmc-20apmz, p-tolyloxy-acetic acid
PubChem CID 70329
IUPAC Name 2-(4-methylphenoxy)acetic acid
SMILES CC1=CC=C(C=C1)OCC(=O)O

Specifications

Melting Point 140°C
Color White
Quantity 25 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylphenoxyacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.