Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-(Methylsulfonyl)acetophenone 98.0+%, TCI America™
SDP

Catalog No. M1678250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1678250G 250 g
M167825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1678250G Supplier TCI America Supplier No. M1678250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10297-73-1
Molecular Formula C9H10O3S
Molecular Weight (g/mol) 198.236
InChI Key KAVZYDHKJNABPC-UHFFFAOYSA-N
Synonym 4'-methylsulfonyl acetophenone, 1-4-methylsulfonyl phenyl ethanone, 4-methylsulfonylacetophenone, 4-methylsulfonyl acetophenone, 4-acetylphenyl methyl sulphone, 1-4-methylsulfonylphenyl ethanone, 4-methylsulphonylacetophenone, 1-4-methanesulfonyl-phenyl-ethanone, p-methylsulfonylacetophenone, 4-methanesulfonylacetophenone
PubChem CID 82529
IUPAC Name 1-(4-methylsulfonylphenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)S(=O)(=O)C

Specifications

Melting Point 129°C
Color White-Yellow
Quantity 250 g
Formula Weight 198.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-(Methylsulfonyl)acetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.