Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(4-Methylsulfonylaminomethyl)benzeneboronic acid, 98%, Thermo Scientific™

Catalog No. AAH5267603
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5267603 1 g
AAH52676MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5267603 Supplier Thermo Scientific Chemicals Supplier No. H5267603

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(4-Methylsulfonylaminomethyl)benzeneboronic acid, is used as a pharmaceutical intermediate.

Solubility
Insoluble in water. Soluble in organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850568-38-6
Molecular Formula C8H12BNO4S
Molecular Weight (g/mol) 229.057
MDL Number MFCD06659816
InChI Key DFNWPOUFTMAIGJ-UHFFFAOYSA-N
Synonym 4-methylsulfonamidomethyl phenyl boronic acid, 4-methanesulfonylaminomethylphenyl boronic acid, 4-methylsulphonylamino methyl benzeneboronic acid, 4-methanesulfonylaminomethyl phenylboronic acid, 4-methanesulfonamidomethyl phenylboronic acid, 4-methylsulfonylaminomethyl benzeneboronic acid, 4-methylsulfonamidomethyl phenylboronic acid, 4-methylsulfonamidomethyl phenyl boronicacid, 4-methanesulfonylaminomethyl phenyl boronic acid
PubChem CID 22309452
IUPAC Name [4-(methanesulfonamidomethyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)CNS(=O)(=O)C)(O)O

Specifications

Melting Point 237°C to 239°C
Quantity 1 g
Formula Weight 229.06
Percent Purity 98%
Chemical Name or Material (4-Methylsulfonylaminomethyl)benzeneboronic acid
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.