Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Methylthiazole, 99%

Click to view available options
Quantity:
5g
25g
100g
693-95-8
C4H5NS
99.151
MFCD00005340
QMHIMXFNBOYPND-UHFFFAOYSA-N
4-methylthiazole,thiazole, 4-methyl,4-methyl thiazole,4-methyl-thiazole,fema no. 3716,4-methyl-5h-1,3-thiazole,4-methyithiazole,methylthiazole, 4,pubchem23683
12748
CHEBI:35626
4-methyl-1,3-thiazole
CC1=CSC=N1
693-95-8
C4H5NS
99.151
MFCD00005340
QMHIMXFNBOYPND-UHFFFAOYSA-N
4-methylthiazole,thiazole, 4-methyl,4-methyl thiazole,4-methyl-thiazole,fema no. 3716,4-methyl-5h-1,3-thiazole,4-methyithiazole,methylthiazole, 4,pubchem23683
12748
CHEBI:35626
4-methyl-1,3-thiazole
CC1=CSC=N1
Refractive Index 1.524
Density 1.093
Boiling Point 133°C to 134°C
Odor Characteristic
Quantity 100g
Flash Point 32°C (89°F)
UN Number UN1993
Beilstein 105228
Formula Weight 99.16
Percent Purity 99%
Chemical Name or Material 4-Methylthiazole
Show More Show Less

GHS H Statement
H301-H226-H315-H319-H335
Toxic if swallowed.
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c

H226-H302-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 211-764-1

RTECSNumber : XJ5096000

TSCA : Yes

Recommended Storage : Ambient temperatures