Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylthio-2-butanone 97.0+%, TCI America™
SDP

Catalog No. M203125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M203125G 25 g
M20315G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M203125G Supplier TCI America Supplier No. M203125G

Chemical Identifiers

CAS 34047-39-7
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.194
MDL Number MFCD00040029
InChI Key DRGHCRKOWMAZAO-UHFFFAOYSA-N
Synonym 4-methylthio-2-butanone, 2-butanone, 4-methylthio, 4-methylthio butan-2-one, unii-5703w1o1ab, 4-methylsulfanyl butan-2-one, potato butanone, fema no. 3375, 5-thiahexane-2-one, acmc-1afub
PubChem CID 61922
IUPAC Name 4-methylsulfanylbutan-2-one
SMILES CC(=O)CCSC

Specifications

Color Yellow
Boiling Point 106°C
Quantity 25 g
Formula Weight 118.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Methylthio-2-butanone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.