Learn More
4-(Methylthio)benzaldehyde, 97%
Description
4-(Methylthio)benzaldehyde is a building block for the synthesis of various pharmaceutical and biologically active compounds. It can be used for the synthesis of sulfur-containing terpyridine ligands. It is the intermediate for the synthesis of a class of pyrrole derivatives showing analgesic/anti-inflammatory activity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-(Methylthio)benzaldehyde is a building block for the synthesis of various pharmaceutical and biologically active compounds. It can be used for the synthesis of sulfur-containing terpyridine ligands. It is the intermediate for the synthesis of a class of pyrrole derivatives showing analgesic/anti-inflammatory activity.
Solubility
Not miscible in water.
Notes
Air sensitive. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 6°C |
| Density | 1.144 |
| Boiling Point | 118°C to 121°C (5 mmHg) |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.649 |
| Quantity | 100 g |
| Beilstein | 636158 |
| Sensitivity | Air Sensitive |
| Solubility Information | Not miscible in water. |
| Formula Weight | 152.22 |
| Show More |
Safety and Handling
| Hazard Category | H302 |
| Precautionary Statement | P264b-P270-P301+P312-P330-P501c |
| EINECSNumber | 222-365-7 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.