Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(Methylthio)benzoic acid, 98%, Thermo Scientific™

Catalog No. AC163460250
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass Bottle

Chemical Identifiers

CAS 13205-48-6
Molecular Formula C8H8O2S
Molecular Weight (g/mol) 168.21
MDL Number MFCD00002555
InChI Key KWHCPERWLHBLOT-UHFFFAOYSA-N
Synonym 4-methylthio benzoic acid, 4-methylsulfanyl benzoic acid, benzoic acid, 4-methylthio, p-methylthio benzoic acid, p-methiobenzoic acid, benzoic acid, p-methylthio, 4-methylthiobenzoicacid, pubchem10665, acmc-1bqaa
PubChem CID 83230
IUPAC Name 4-(methylsulfanyl)benzoic acid
SMILES CSC1=CC=C(C=C1)C(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 192°C to 196°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Linear Formula CH3SC6H4COOH
Quantity 25 g
Beilstein 10, I, 80
Formula Weight 168.22
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material 4-(Methylthio)benzoic acid
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.