Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylumbelliferyl acetate, 99%, Thermo Scientific™

Catalog No. p-7081949
Click to view available options
Quantity:
100 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2747-05-9
ChEBI CHEBI:17763

Specifications

Melting Point 147°C to 152°C
Molecular Formula C12H10O4
Quantity 100 g
MDL Number MFCD00006865
Beilstein 189667
Synonym 7-acetoxy-4-methylcoumarin, 4-methylumbelliferyl acetate, 4-methyl-2-oxo-2h-chromen-7-yl acetate, mu-ac, 2h-1-benzopyran-2-one, 7-acetyloxy-4-methyl, 7-acetyl-4-methylcoumarin, beta-methylumbelliferyl acetate, 7-acetyloxy-4-methyl-2-benzopyrone, 4-methyl-2-oxochromen-7-yl acetate, 4-methyl-7-acetyloxy coumarin
InChI Key HXVZGASCDAGAPS-UHFFFAOYSA-N
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OC(=O)C
IUPAC Name (4-methyl-2-oxochromen-7-yl) acetate
Molecular Weight (g/mol) 218.21
PubChem CID 366
Formula Weight 218.21
Percent Purity 99%
Chemical Name or Material 4-Methylumbelliferyl acetate
Show More Show Less
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-386-6
TSCA No
Recommended Storage Keep cold
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.