Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylumbelliferyl beta-D-Ribofuranoside 98.0+%, TCI America™
SDP

Catalog No. M3027100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M3027100MG 100 mg
M302725MG 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M3027100MG Supplier TCI America Supplier No. M3027100MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 195385-93-4
Molecular Formula C15H16O7
Molecular Weight (g/mol) 308.29
MDL Number MFCD06796941
InChI Key FAGLTVBWEMHJRP-UHFFFAOYNA-N
PubChem CID 18731513
IUPAC Name 7-{[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]oxy}-4-methyl-2H-chromen-2-one
SMILES CC1=CC(=O)OC2=CC(OC3OC(CO)C(O)C3O)=CC=C12

Specifications

Color White
Quantity 100 mg
Formula Weight 308.29
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Methylumbelliferyl beta-D-Ribofuranoside
TSCA No
Recommended Storage Freezer (-20 °C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.