Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
4-Methylumbelliferyl-beta-D-glucopyranoside, 99%
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                500 mg
                            
                        
                            
                                2.5 g
                            
                        
                    
                            
                                Packaging:
                            
                        
                        
                            
                                Glass bottle
                            
                        
                    Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
 
            Specifications
Specifications
| CAS Min % | 98.5 | 
| CAS Max % | 100.0 | 
| Melting Point | 210.0°C to 212.0°C | 
| Color | White | 
| Infrared Spectrum | Authentic | 
| Assay Percent Range | 99% | 
| Packaging | Glass bottle | 
| Quantity | 500 mg | 
| Beilstein | 18, V,1, 443 | 
| Specific Rotation Condition | −99.00 (20.00°C c=0.3,water) | 
| Show More | 
Safety and Handling
| Hazard Category | MOISTURE SENSITIVE | 
| EINECSNumber | 242-736-7 | 
RUO – Research Use Only
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                                