Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Methylumbelliferyl phosphate, 99%

Catalog No. p-7043189
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 3368-04-5 | C10H9O6P | 256.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3368-04-5
MDL Number MFCD00016969

Specifications

Melting Point 214°C to 216°C
Color White
Molecular Formula C10H9O6P
Quantity 250 mg
Beilstein 1687229
Synonym 4-methylumbelliferyl phosphate, 4-methyl-2-oxo-2h-chromen-7-yl dihydrogen phosphate, 4-mup, 2h-1-benzopyran-2-one, 4-methyl-7-phosphonooxy, phosphoric acid 4-methylumbelliferyl ester, 4-methylumbelliferylphosphate, free acid, 4-methyl-2-oxochromen-7-yl oxyphosphonic acid, 5,5-dinitrobarbutyric acid, 4-methylumbelliferone phosphate, 4-methylumbelliferyl phosphate, free acid
Solubility Information Soluble in water
InChI Key BCHIXGBGRHLSBE-UHFFFAOYSA-N
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)OP(=O)(O)O
IUPAC Name (4-methyl-2-oxochromen-7-yl) dihydrogen phosphate
Molecular Weight (g/mol) 256.15
PubChem CID 65118
Formula Weight 256.15
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 4-Methylumbelliferyl phosphate
Show More Show Less
EINECSNumber 222-137-7
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.