Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Morpholinophenylboronic acid, 97%, Thermo Scientific™

Catalog No. CC17412DA
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC17412DA 1 g Amber Glass Bottle
CC17412CB 250 mg Amber Glass Bottle
CC17412DE 5 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC17412DA Supplier Thermo Scientific Chemicals Supplier No. CC17412DA
Add to Cart

Chemical Identifiers

CAS 186498-02-2
Molecular Formula C10H14BNO3
Molecular Weight (g/mol) 207.04
MDL Number MFCD03095169
InChI Key WHDIUBHAKZDSJL-UHFFFAOYSA-N
Synonym 4-morpholinophenylboronic acid, 4-morpholinophenyl boronic acid, 4-morpholino phenylboronic acid, 4-morpholin-4-yl-phenylboronic acid, 4-morpholin-4-ylphenyl boronic acid, 4-4-boronophenyl morpholine, 4-morpholino phenyl boronic acid, 4-morpholin-4-yl phenylboronic acid, boronic acid, 4-4-morpholinyl phenyl, boronic acid, b-4-4-morpholinyl phenyl
PubChem CID 2795359
IUPAC Name (4-morpholin-4-ylphenyl)boronic acid
SMILES OB(O)C1=CC=C(C=C1)N1CCOCC1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 207.04
Percent Purity 97%
Chemical Name or Material 4-Morpholinophenylboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.