Alfa Aesar™ 4-n-Butoxyphenol, 98%

Manufacturer: Alfa Aesar™ A1207606

Quantity 5g

 View more versions of this product

Catalog No. AAA1207606

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 122-94-1
    Molecular Formula C10H14O2
    Formula Weight 166.22
    MDL Number MFCD00002336
    Synonym Hydroquinone mono-n-butyl ether
    Chemical Name or Material 4-n-Butoxyphenol

    Specifications

    Formula Weight 166.22
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 122-94-1
    Quantity 5g
    Molecular Formula C10H14O2
    Synonym Hydroquinone mono-n-butyl ether
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    MDL Number MFCD00002336
    Melting Point 63° to 66°C
    TSCA TSCA
    Chemical Name or Material 4-n-Butoxyphenol
    Assay 98%

    Description

    Applications
    It is an intermediate of liquid crystals.

    Solubility
    Soluble in acetone and ethanol.

    Notes
    Store in cool place. Keep away from oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit