Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-n-Dodecylaniline, 95%

Click to view available options
Quantity:
1g
5g
104-42-7
C18H31N
261.453
MFCD00007919
KLPPPIIIEMUEGP-UHFFFAOYSA-N
p-dodecylaniline,benzenamine, 4-dodecyl,p-dodecyl aniline,4-dodecylphenylamine,4-n-dodecylamine,p-n-dodecylaniline,paradodecyl aniline,acmc-20amjq,4-dodecylaniline,ksc180i8p
7701
4-dodecylaniline
CCCCCCCCCCCCC1=CC=C(C=C1)N
104-42-7
C18H31N
261.453
MFCD00007919
KLPPPIIIEMUEGP-UHFFFAOYSA-N
p-dodecylaniline,benzenamine, 4-dodecyl,p-dodecyl aniline,4-dodecylphenylamine,4-n-dodecylamine,p-n-dodecylaniline,paradodecyl aniline,acmc-20amjq,4-dodecylaniline,ksc180i8p
7701
4-dodecylaniline
CCCCCCCCCCCCC1=CC=C(C=C1)N
Applications
4-Dodecylaniline has been used: in the synthesis of sodium 4-dodecylphenylazosulfonate, to investigate the water solubilization of single-walled carbon nanotubes (SWNTs), in proton transfer step during molecular oxygen (O2) reduction by decamethylferrocene (DMFc).

Notes
Store in a cool, dry conditions in a well sealed container. Store away from oxidizing agents.
Quantity 1g
Melting Point 35°C to 39°C
Boiling Point 220°C to 221°C (15mmHg)
UN Number UN3077
Beilstein 2646305
Formula Weight 261.45
Physical Form Solid
Percent Purity 95%
Chemical Name or Material 4-n-Dodecylaniline

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

TSCA : TSCA