Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-n-Dodecyloxybenzoic acid, 98%

Click to view available options
Quantity:
5g
25g
2312-15-4
C19H30O3
306.446
MFCD00002543
ALQLYJHDBAKLBB-UHFFFAOYSA-N
4-dodecyloxy benzoic acid,p-dodecyloxybenzoic acid,4-n-dodecyloxybenzoic acid,4-dodecyloxybenzoic acid,benzoic acid, 4-dodecyloxy,benzoic acid, p-dodecyloxy,p-dodecyloxy benzoic acid,4-dodecyloxy benzoicacid,pubchem2685,acmc-1crse
75330
4-dodecoxybenzoic acid
CCCCCCCCCCCCOC1=CC=C(C=C1)C(=O)O
2312-15-4
C19H30O3
306.446
MFCD00002543
ALQLYJHDBAKLBB-UHFFFAOYSA-N
4-dodecyloxy benzoic acid,p-dodecyloxybenzoic acid,4-n-dodecyloxybenzoic acid,4-dodecyloxybenzoic acid,benzoic acid, 4-dodecyloxy,benzoic acid, p-dodecyloxy,p-dodecyloxy benzoic acid,4-dodecyloxy benzoicacid,pubchem2685,acmc-1crse
75330
4-dodecoxybenzoic acid
CCCCCCCCCCCCOC1=CC=C(C=C1)C(=O)O
Applications
4-n-Dodecyloxybenzoic acid is used as an organic chemical synthesis intermediate.

Solubility
Insoluble in water. Soluble in N,N-Dimethylformamide.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Melting Point 132°C to 137°C
Quantity 25g
Beilstein 2215914
Formula Weight 306.45
Percent Purity 98%
Chemical Name or Material 4-n-Dodecyloxybenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning