Learn More
4-n-Hexylaniline, 98%
Description
4-Hexylaniline is used in the fabrication of RP/ion-exchange, mixed-mode, monolithic materials for capillary LC. The product is dye stuff intermediate, yet it may use to manufacture the medicine
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
4-Hexylaniline is used in the fabrication of RP/ion-exchange, mixed-mode, monolithic materials for capillary LC. The product is dye stuff intermediate, yet it may use to manufacture the medicine
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids.

Specifications
Specifications
Density | 0.919 |
Boiling Point | 283°C to 285°C |
Flash Point | >110°C (230°F) |
Refractive Index | 1.525 |
Quantity | 25 g |
Sensitivity | Light sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 177.29 |
Percent Purity | 98% |
Chemical Name or Material | 4-n-Hexylaniline |
Safety and Handling
Hazard Category | H302-H315-H319 |
Hazard Statement | GHS H Statement H302-H315-H319 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c |
EINECSNumber | 251-409-8 |
RTECSNumber | BY2305000 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.