Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-n-Hexylbiphenyl-4-carboxylic acid, 99%

Catalog No. p-7026096
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 59662-48-5 | C19H33O2 | 293.47 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59662-48-5
Molecular Formula C19H33O2
Molecular Weight (g/mol) 293.47
MDL Number MFCD00143232
InChI Key NWHAWRQJZXUDLX-UHFFFAOYSA-M
Synonym 4-4-hexylphenyl benzoic acid, 4-hexyl-4'-biphenylcarboxylic acid, 4-hexyl-4'-carboxybiphenyl, 4-n-hexylbiphenyl-4'-carboxylic acid, 4'-hexyl-1,1'-biphenyl-4-carboxylic acid, pubchem9069, acmc-209mez, 4'-hexylbiphenyl-4-carboxylic acid, 4-hexylbiphenyl-4'-carboxylic acid, 4'-hexyl-biphenyl-4-carboxylic acid
PubChem CID 13554588
IUPAC Name 4-(4-hexylphenyl)benzoic acid
SMILES CCCCCCC1CCC(CC1)C1CCC(CC1)C([O-])=O

Specifications

Melting Point 222°C to 224°C
Quantity 1 g
Formula Weight 282.38
Percent Purity 99%
Chemical Name or Material 4'-n-Hexylbiphenyl-4-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.