Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-n-Nonylphenol, 98+%

Catalog No. aaa1560903
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 104-40-5 | C15H24O | 220.356 g/mol

4-n-Nonylphenol is used in manufacturing antioxidants, lubricating oil additives, laundry and dish detergents, emulsifiers, and solubilizers. Also be used to produce tris(4-nonyl-phenyl) phosphite (TNPP).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-n-Nonylphenol is used in manufacturing antioxidants, lubricating oil additives, laundry and dish detergents, emulsifiers, and solubilizers. Also be used to produce tris(4-nonyl-phenyl) phosphite (TNPP).

Solubility
Sparingly Soluble (0.020 g/L) at 25°C.

Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-40-5
Molecular Formula C15H24O
Molecular Weight (g/mol) 220.356
MDL Number MFCD00002396
InChI Key IGFHQQFPSIBGKE-UHFFFAOYSA-N
Synonym 4-n-nonylphenol, p-nonylphenol, phenol, 4-nonyl, para-nonylphenol, p-n-nonylphenol, phenol, nonyl, phenol, p-nonyl, nonylphenol mixed, phenol, nonyl derivs., 4-nonyl phenol
PubChem CID 1752
ChEBI CHEBI:34440
IUPAC Name 4-nonylphenol
SMILES CCCCCCCCCC1=CC=C(C=C1)O

Specifications

Melting Point 43°C to 46°C
Density 0.937
Boiling Point 127°C to 130°C (27 mmHg)
Flash Point 150°C (302°F)
Refractive Index 1.511
Quantity 1 g
UN Number UN2430
Beilstein 2047450
Solubility Information Sparingly Soluble (0.020g/L) at 25°C.
Formula Weight 220.36
Percent Purity ≥98%
Chemical Name or Material 4-n-Nonylphenol
Show More Show Less
Hazard Category H302-H314-H335-H361fd
Hazard Statement GHS H Statement
H314-H318-H361-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, SOLID, N.O.S.
EINECSNumber 203-199-4
RTECSNumber SM5630000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.