Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-n-Octylbiphenyl-4-carbonitrile, 99%, Thermo Scientific Chemicals

Catalog No. p-7028903
Click to view available options
Quantity:
1 g

The thermobarometry of thermotropic liquid crystals is feasible by the metal tube Berthelot method, and its extension in the negative-pressure range will be interesting for the study of organic compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The thermobarometry of thermotropic liquid crystals is feasible by the metal tube Berthelot method, and its extension in the negative-pressure range will be interesting for the study of organic compounds.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52709-84-9
Molecular Formula C21H25N
Molecular Weight (g/mol) 291.44
MDL Number MFCD00075146
InChI Key CSQPODPWWMOTIY-UHFFFAOYSA-N
Synonym 4-cyano-4'-octylbiphenyl, 4'-octyl-1,1'-biphenyl-4-carbonitrile, 4-4-octylphenyl benzonitrile, 1,1'-biphenyl-4-carbonitrile, 4'-octyl, 4-octylcyanodiphenyl, k 24 liquid crystal, cb 8, 4'-octyl-4-biphenylcarbonitrile, 4'-octyl 1,1'-biphenyl-4-carbonitrile, 4-octyl-4'-cyanobiphenyl
PubChem CID 104289
IUPAC Name 4-(4-octylphenyl)benzonitrile
SMILES CCCCCCCCC1=CC=C(C=C1)C1=CC=C(C=C1)C#N

Specifications

Melting Point 20°C to 22°C
Flash Point >110°C (230°F)
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 291.43
Percent Purity 99%
Chemical Name or Material 4'-n-Octylbiphenyl-4-carbonitrile
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 258-120-6
RTECSNumber DV1764400
TSCA Yes

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.