Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-n-Octylbiphenyl, 97%, Thermo Scientific™

Catalog No. AAB2501003
Click to view available options
Quantity:
1g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7116-97-4
Molecular Formula C20H26
Quantity 1g
MDL Number MFCD00102113
Synonym 4-octylbiphenyl, 4-octyl-1,1'-biphenyl, 4-n-octylbiphenyl, 4-octyldiphenyl, 1,1'-biphenyl, 4-octyl, biphenyl, 4-octyl-6ci,7ci,8ci, 4-octyl-1-phenylbenzene, 4-05-00-02003 beilstein handbook reference
InChI Key KCLBLDDJJJNGBC-UHFFFAOYSA-N
SMILES CCCCCCCCC1=CC=C(C=C1)C2=CC=CC=C2
IUPAC Name 1-octyl-4-phenylbenzene
Molecular Weight (g/mol) 266.428
PubChem CID 201918
Formula Weight 266.43
Percent Purity 97%
Chemical Name or Material 4-n-Octylbiphenyl
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.