Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-[N-(p-Methoxybenzylidene)amino]-phenyl Acetate, Thermo Scientific™

Catalog No. AC413800250
Click to view available options
Quantity:
25 g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10484-13-6
Molecular Formula C16H15NO3
Molecular Weight (g/mol) 269.30
MDL Number MFCD00041911
InChI Key HOYWVKUPOCFKOT-UHFFFAOYSA-N
Synonym apapa, n-4-methoxybenzylidene-4-acetoxyaniline, 4'-methoxybenzylidene-4-acetoxyaniline, p-anisylidene-p-aminophenyl acetate, p-n-anisylideneamino phenyl acetate, 4-methoxybenzylidene-4'-aminophenyl acetate, p-p-methoxybenzylidene amino phenyl acetate, n-p-methoxybenzylidene-p-aminophenyl acetate, anisylidene p-aminophenylacetate, p-anisylideneamino phenyl acetate
PubChem CID 97495
IUPAC Name 4-{[(4-methoxyphenyl)methylidene]amino}phenyl acetate
SMILES COC1=CC=C(C=NC2=CC=C(OC(C)=O)C=C2)C=C1

Specifications

Quantity 25 g
Formula Weight 269.29
Chemical Name or Material 4-[N-(p-Methoxybenzylidene)amino]-phenyl acetate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.