Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-n-Pentylaniline, 98%

Catalog No. p-7029133
Click to view available options
Quantity:
5 g
25 g

CAS: 33228-44-3 | C11H17N | 163.264 g/mol

4-Pentylaniline is used in the synthesis of 4-[(4?-pentyl)azo]phenol (5Az-OH) by diazo-coupling reaction with phenol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Pentylaniline is used in the synthesis of 4-[(4′-pentyl)azo]phenol (5Az-OH) by diazo-coupling reaction with phenol

Solubility
Sparingly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33228-44-3
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.264
MDL Number MFCD00007926
InChI Key DGFTWBUZRHAHTH-UHFFFAOYSA-N
Synonym benzenamine, 4-pentyl, 4-amylaniline, p-pentylaniline, p-amylaniline, unii-8roj0g9hiu, p-n-pentylaniline, 4-n-pentylaniline, 4-pentylphenylamine, 8roj0g9hiu, p-n-amylaniline
PubChem CID 93162
IUPAC Name 4-pentylaniline
SMILES CCCCCC1=CC=C(C=C1)N

Specifications

Density 0.919
Boiling Point 130°C
Flash Point >110°C (230°F)
Refractive Index 1.53
Quantity 5 g
Solubility Information Sparingly soluble in water.
Formula Weight 163.26
Percent Purity 98%
Chemical Name or Material 4-n-Pentylaniline
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 251-409-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.