Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-n-Pentylphenol, 98%

Catalog No. AAA1151414
Click to view available options
Quantity:
5 g
25 g

CAS: 14938-35-3 | C11H16O | 164.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14938-35-3
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.25
MDL Number MFCD00020211
InChI Key ZNPSUQQXTRRSBM-UHFFFAOYSA-N
Synonym 4-n-pentylphenol, 4-amylphenol, phenol, 4-pentyl, 4-n-amylphenol, p-pentylphenol, p-amylphenol, phenol, p-pentyl, 4-pentyl-phenol, amyl p-hydroxybenzene, p-n-amylphenol
PubChem CID 26975
ChEBI CHEBI:34441
IUPAC Name 4-pentylphenol
SMILES CCCCCC1=CC=C(O)C=C1

Specifications

Melting Point 24°C
Density 0.96
Boiling Point 342°C
Flash Point 133°C (271°F)
Refractive Index 1.5145
Quantity 25 g
UN Number UN2430
Beilstein 1364424
Formula Weight 164.25
Percent Purity 98%
Chemical Name or Material 4-n-Pentylphenol
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, SOLID, N.O.S.
EINECSNumber 239-015-4
RTECSNumber SM6750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.