Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-n-Propylphenylacetylene, 97%

Click to view available options
Quantity:
5g
25g
100g
62452-73-7
C11H12
144.217
MFCD00173886
UVFFOABHOIMLNB-UHFFFAOYSA-N
4-propylphenylacetylene,1-eth-1-ynyl-4-propylbenzene,4-n-propylphenylacetylene,1-ethynyl-4-propyl-benzene,benzene, 1-ethynyl-4-propyl,pubchem10973,4-n-propylphenyl ethine,4-propyl phenylacetylene,4-ethynyl-1-propylbenzene,4-propyl-1-ethynylbenzene
2775133
1-ethynyl-4-propylbenzene
CCCC1=CC=C(C=C1)C#C
62452-73-7
C11H12
144.217
MFCD00173886
UVFFOABHOIMLNB-UHFFFAOYSA-N
4-propylphenylacetylene,1-eth-1-ynyl-4-propylbenzene,4-n-propylphenylacetylene,1-ethynyl-4-propyl-benzene,benzene, 1-ethynyl-4-propyl,pubchem10973,4-n-propylphenyl ethine,4-propyl phenylacetylene,4-ethynyl-1-propylbenzene,4-propyl-1-ethynylbenzene
2775133
1-ethynyl-4-propylbenzene
CCCC1=CC=C(C=C1)C#C
Quantity 25g
Flash Point 67°C (153°F)
Boiling Point 205°C
Formula Weight 144.22
Percent Purity 97%
Chemical Name or Material 4-n-Propylphenylacetylene