Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-n-Propylpiperidine, 95%

Catalog No. AAH2584803
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2584803 1 g
AAH25848MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2584803 Supplier Thermo Scientific Chemicals Supplier No. H2584803
Add to Cart

CAS: 22398-09-0 | C8H17N | 127.231 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22398-09-0
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.231
MDL Number MFCD00023151
InChI Key RQGBFVLTFYRYKB-UHFFFAOYSA-N
Synonym 4-n-propylpiperidine, piperidine, 4-propyl, 4-propyl-piperidine, pubchem2452, 4-prop-1-yl piperidine, 5-20-04-00208 beilstein handbook reference
PubChem CID 31169
IUPAC Name 4-propylpiperidine
SMILES CCCC1CCNCC1

Specifications

Density 0.864
Boiling Point 178°C to 180°C
Flash Point 48°C (118°F)
Odor Amine-like
Refractive Index 1.453
Quantity 1 g
UN Number UN2733
Sensitivity Air Sensitive
Formula Weight 127.23
Percent Purity 95%
Chemical Name or Material 4-n-Propylpiperidine
Show More Show Less
Hazard Category H226-H314-H335
Hazard Statement GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 244-956-9
RTECSNumber TN3740005
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.