Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Nitro-4-dimethylaminoazobenzene 98.0+%, TCI America™
SDP

Catalog No. N019225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N019225G 25 g
N01925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N019225G Supplier TCI America Supplier No. N019225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2491-74-9
Molecular Formula C14H14N4O2
Molecular Weight (g/mol) 270.29
MDL Number MFCD00059834
InChI Key LSFRFLVWCKLQTO-UHFFFAOYSA-N
PubChem CID 17225
IUPAC Name N,N-dimethyl-4-[2-(4-nitrophenyl)diazen-1-yl]aniline
SMILES CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 229°C
Color Purple-Red
Quantity 25 g
Synonym 4-Dimethylamino-4′-nitroazobenzene
Formula Weight 270.29
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Nitro-4-dimethylaminoazobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.