Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitro-N-(n-propyl)benzylamine hydrochloride, 99%, Thermo Scientific™

Catalog No. AAA1930803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1930803 1 g
AAA1930806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1930803 Supplier Thermo Scientific Chemicals Supplier No. A1930803

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Nitro-N-propylbenzylamine hydrochloride was used as a derivatization reagent for the extraction of amine in the determination of trace amounts of diisocyanate monomers in contaminated air by an electroanalytical method.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68133-98-2
Molecular Formula C10H15ClN2O2
Molecular Weight (g/mol) 230.692
MDL Number MFCD00012845
InChI Key LWISLZIFEARHJI-UHFFFAOYSA-N
Synonym 4-nitro-n-propylbenzylamine hydrochloride, n-4-nitrobenzyl-n-propylamine hydrochloride, benzenemethanamine, 4-nitro-n-propyl-, monohydrochloride, n-4-nitrobenzyl propylamine hydrochloride, benzenemethanamine, 4-nitro-n-propyl-, hydrochloride 1:1, 4-nitrophenyl methyl propyl amine hydrochloride, n-4-nitrophenyl methyl propan-1-amine hydrochloride, 4-nitrophenyl methyl propylamine, chloride, p-nitro-n-propylbenzylamine hydrochloride, pnbpa
PubChem CID 109563
IUPAC Name N-[(4-nitrophenyl)methyl]propan-1-amine;hydrochloride
SMILES CCCNCC1=CC=C(C=C1)[N+](=O)[O-].Cl

Specifications

Melting Point 228°C to 234°C
Quantity 1 g
Beilstein 3714783
Solubility Information Soluble in water.
Formula Weight 230.7
Percent Purity 99%
Chemical Name or Material 4-Nitro-N-(n-propyl)benzylamine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 268-721-5
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.