Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Potassium thiocyanate, 99%, ACS reagent
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator

Specifications
Specifications
Melting Point | 170.0°C to 179.0°C |
Color | Colorless or White |
pH | 5.3 to 8.7 (5% soln. at 25°C) |
Physical Form | Crystals |
Quantity | 2.5 kg |
Assay Percent Range | 99% |
Linear Formula | KSCN |
Fieser | 01,954 |
Merck Index | 15, 7807 |
Solubility Information | Solubility in water: 2170g/L (20°C). Other solubilities: soluble in alcohol and acetone |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Warning |
Hazard Statement | GHS H Statement: Harmful to aquatic life with long lasting effects. Harmful if swallowed. Harmful if inhaled. Harmful in contact with skin. Contact with acids liberates very toxic gas. |
Precautionary Statement | GHS P Statement: Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Avoid release to the environment. Keep container tightly closed. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification. |
EINECSNumber | 206-370-1 |
RTECSNumber | XL1925000 |
TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction