Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Nitroacetophenone, 98%

Catalog No. aaa1387718
Click to view available options
Quantity:
50 g
250 g

CAS: 100-19-6 | C8H7NO3 | 165.15 g/mol

4'-Nitroacetophenone is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4′-Nitroacetophenone is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent, strong alkalis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-19-6
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.15
MDL Number MFCD00007355
InChI Key YQYGPGKTNQNXMH-UHFFFAOYSA-N
Synonym p-nitroacetophenone, 4'-nitroacetophenone, 4-nitroacetophenone, 1-4-nitrophenyl ethanone, p-acetylnitrobenzene, paranitroacetophenone, p-nitrophenyl methyl ketone, 1-4-nitrophenyl ethan-1-one, ethanone, 1-4-nitrophenyl, pnap
PubChem CID 7487
ChEBI CHEBI:28735
IUPAC Name 1-(4-nitrophenyl)ethanone
SMILES CC(=O)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 77°C to 81°C
Boiling Point 201°C to 202°C
Flash Point 149°C (300°F)
Assay Percent Range 98%
Quantity 50 g
Beilstein 607777
Solubility Information Insoluble in water.
Formula Weight 165.15
Percent Purity 98%
Chemical Name or Material 4'-Nitroacetophenone
Hazard Category H302-H319-H500
Hazard Statement GHS H Statement
H301
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
EINECSNumber 202-827-4
RTECSNumber AM9627000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.