Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitroaniline, 99%

Catalog No. AC128371000
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 100-01-6 | C6H6N2O2 | 138.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-01-6
Molecular Formula C6H6N2O2
Molecular Weight (g/mol) 138.13
MDL Number MFCD00007858
InChI Key TYMLOMAKGOJONV-UHFFFAOYSA-N
Synonym p-nitroaniline, p-nitraniline, 4-nitrobenzenamine, p-aminonitrobenzene, 4-nitraniline, benzenamine, 4-nitro, p-nitrophenylamine, 1-amino-4-nitrobenzene, aniline, p-nitro, developer p
PubChem CID 7475
ChEBI CHEBI:17064
IUPAC Name 4-nitroaniline
SMILES NC1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 146.0°C to 151.0°C
Color Yellow to Brown
Boiling Point 332.0°C
Flash Point 199°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4NH2
Quantity 100 g
Beilstein 12,711
Merck Index 15,667
Solubility Information Solubility in water: 0.8g/L (20°C). Other solubilities: soluble in alcohol,ether,benzene and methanol,1 g soluble in 25 mL alcohol,1 g soluble in 30 mL ether
Formula Weight 138.13
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material 4-Nitroaniline
Show More Show Less
missing translation for 'healthHazard1' GHS Signal Word: Danger
missing translation for 'healthHazard2' GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.
missing translation for 'healthHazard3' GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breat
EINECSNumber 202-810-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.