Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitroaniline, 99%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
2.5kg
Packaging:
Glass bottle
Plastic Bottle
100-01-6
C6H6N2O2
138.13
TYMLOMAKGOJONV-UHFFFAOYSA-N
p-nitroaniline,p-nitraniline,4-nitrobenzenamine,p-aminonitrobenzene,4-nitraniline,benzenamine, 4-nitro,p-nitrophenylamine,1-amino-4-nitrobenzene,aniline, p-nitro,developer p
7475
CHEBI:17064
4-nitroaniline
C1=CC(=CC=C1N)[N+](=O)[O-]
100-01-6
C6H6N2O2
138.13
TYMLOMAKGOJONV-UHFFFAOYSA-N
p-nitroaniline,p-nitraniline,4-nitrobenzenamine,p-aminonitrobenzene,4-nitraniline,benzenamine, 4-nitro,p-nitrophenylamine,1-amino-4-nitrobenzene,aniline, p-nitro,developer p
7475
CHEBI:17064
4-nitroaniline
C1=CC(=CC=C1N)[N+](=O)[O-]
Indicator
Flash Point 199°C
Quantity 500g
Infrared Spectrum Authentic
Color Yellow to Brown
Boiling Point 332.0°C
Packaging Glass bottle
Melting Point 146.0°C to 151.0°C
Assay Percent Range 99%
Linear Formula O2NC6H4NH2
Beilstein 12,711
Merck Index 15,667
Solubility Information Solubility in water: 0.8g/L (20°C). Other solubilities: soluble in alcohol,ether,benzene and methanol,1 g soluble in 25 mL alcohol,1 g soluble in 30 mL ether
Formula Weight 138.13
Physical Form Crystalline Powder, Crystals or Flakes
Percent Purity 99%
Chemical Name or Material 4-Nitroaniline
Show More Show Less