Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrobenzaldehyde, 99%

Catalog No. ac156990250
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle

CAS: 555-16-8 | C7H5NO3 | 151.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 555-16-8
Molecular Formula C7H5NO3
Molecular Weight (g/mol) 151.12
MDL Number MFCD00007346
InChI Key BXRFQSNOROATLV-UHFFFAOYSA-N
Synonym p-nitrobenzaldehyde, benzaldehyde, 4-nitro, p-formylnitrobenzene, benzaldehyde, p-nitro, 4-nitrobenzaldehydde, 4-nitro-benzaldehyde, 4-formylnitrobenzene, p-nitro benzaldehyde, 4-nitro benzaldehyde, ccris 1675
PubChem CID 541
ChEBI CHEBI:66926
IUPAC Name 4-nitrobenzaldehyde
SMILES [O-][N+](=O)C1=CC=C(C=O)C=C1

Specifications

Melting Point 105.0°C to 106.0°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula O2NC6H4CHO
Quantity 25 g
Beilstein 07,256
Merck Index 14,6587
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in glacial acetic acid,soluble in alcohol and benzene,slightly soluble in ether
Formula Weight 151.12
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrobenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
EINECSNumber 209-084-5

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.