Alfa Aesar™ 4-Nitrobenzenediazonium tetrafluoroborate, 97%

Manufacturer: Alfa Aesar™ H5531503

Quantity 1g

 View more versions of this product

Catalog No. AAH5531503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 456-27-9
    Molecular Formula C6H4BF4N3O2
    Formula Weight 236.92
    MDL Number MFCD00012005
    UN Number UN2923
    Beilstein 3599869
    Synonym Fast Red GG Salt; C.I. 37035
    Chemical Name or Material 4-Nitrobenzenediazonium tetrafluoroborate

    Specifications

    Formula Weight 236.92
    TSCA TSCA
    MDL Number MFCD00012005
    UN Number UN2923
    Synonym Fast Red GG Salt; C.I. 37035
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
    RTECS Number CZ1723000
    Melting Point 144° to 148°C
    Molecular Formula C6H4BF4N3O2
    CAS 456-27-9
    Beilstein 3599869
    Health Hazard 2 GHS H Statement
    H314-H318-H302-H312-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 1g
    Chemical Name or Material 4-Nitrobenzenediazonium tetrafluoroborate
    Assay 97%

    Description

    Applications
    4-Nitrobenzenediazonium tetrafluoroborate is widely used as a standard reagent in organic synthesis. It is used as a dye for enterochromaffin cells. It is used for the spectrophotometric determination of common aromatic diisocyanates [toluene-2,4-diyl diisocyanate and 4,4 ft.-methylenebis(phenyl isocyanate)] and gallic acid. It is also used for tissue staining and determination of trace phenols.

    Solubility
    Soluble in acetonitrile and dimethyl sulfoxide.

    Notes
    Moisture sensitive. Store in a cool place. Keep the container tightly closed. Incompatible with strong bases, strong oxidizing agents and moisture.

    GHS H Statement
    H314-H318-H302-H312-H332
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit