Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrobenzoic acid, 99%

Catalog No. p-7022118
Click to view available options
Quantity:
250 g
1000 g
5000 g

CAS: 62-23-7 | C7H5NO4 | 167.12 g/mol

4-Nitrobenzoic acid used as an intermediate for bulk drugs especially in the manufacturing of folic acid and in the manufacturing of PABA, DABA & dye intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzoic acid used as an intermediate for bulk drugs especially in the manufacturing of folic acid and in the manufacturing of PABA, DABA & dye intermediate.

Solubility
Very slightly soluble in cold water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-23-7
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007352
InChI Key OTLNPYWUJOZPPA-UHFFFAOYSA-N
Synonym 4-nitrobenzoic acid, p-nitrobenzoic acid, benzoic acid, 4-nitro, nitrodracylic acid, 4-nitrodracylic acid, 1-carboxy-4-nitrobenzene, benzoic acid, p-nitro, p-nitrobenzoicacid, p-nitrobenzenecarboxylic acid, kyselina p-nitrobenzoova
PubChem CID 6108
ChEBI CHEBI:262350
SMILES C1=CC(=CC=C1C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 239°C to 241°C
Density 1.61
Quantity 250 g
Beilstein 973593
Merck Index 14,6589
Solubility Information Very slightly soluble in cold water.
Formula Weight 167.12
Percent Purity 99%
Chemical Name or Material 4-Nitrobenzoic acid
Hazard Category H302-H319
Hazard Statement GHS H Statement
H301-H351-H315-H319-H335
Toxic if swallowed.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c
EINECSNumber 200-526-2
RTECSNumber DH5075000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.