Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrobenzonitrile, 97%

Catalog No. p-173007
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 619-72-7 | C7H4N2O2 | 148.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-72-7
Molecular Formula C7H4N2O2
Molecular Weight (g/mol) 148.12
MDL Number MFCD00007279
InChI Key NKJIFDNZPGLLSH-UHFFFAOYSA-N
Synonym benzonitrile, 4-nitro, 4-cyanonitrobenzene, p-nitrobenzonitrile, p-cyanonitrobenzene, benzonitrile, p-nitro, 4-nitrobenzenecarbonitrile, 1-nitro-4-cyanobenzene, 4-nitro-benzonitrile, ccris 2328, 4-nitrobenzonitril
PubChem CID 12090
IUPAC Name 4-nitrobenzonitrile
SMILES [O-][N+](=O)C1=CC=C(C=C1)C#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 144.0°C to 149.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula O2NC6H4CN
Quantity 25 g
Beilstein 09,397
Solubility Information Solubility in water: sparingly soluble
Formula Weight 148.12
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Nitrobenzonitrile, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Fatal if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash w
EINECSNumber 210-610-0
RTECSNumber DI4903500
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.