Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrobenzyl Chloroformate 97%, ACROS Organics™

Click to view available options
Quantity:
25g
4457-32-3
C8H6ClNO4
215.589
MHSGOABISYIYKP-UHFFFAOYSA-N
4-nitrobenzyl chloroformate,4-nitrobenzyl carbonochloridate,carbonochloridic acid, 4-nitrophenyl methyl ester,4-nitrobenzyl chlorocarbonate,p-nitrobenzyl chloroformate,4-nitrocarbobenzoxychloride,4-nitrophenyl methyl chloroformate,p-nitrobenzyloxycarbonyl chloride,4-nitrobenzyloxycarbonyl chloride,chloroformic acid p-nitrobenzyl ester
78205
(4-nitrophenyl)methyl carbonochloridate
C1=CC(=CC=C1COC(=O)Cl)[N+](=O)[O-]
4457-32-3
C8H6ClNO4
215.589
MHSGOABISYIYKP-UHFFFAOYSA-N
4-nitrobenzyl chloroformate,4-nitrobenzyl carbonochloridate,carbonochloridic acid, 4-nitrophenyl methyl ester,4-nitrobenzyl chlorocarbonate,p-nitrobenzyl chloroformate,4-nitrocarbobenzoxychloride,4-nitrophenyl methyl chloroformate,p-nitrobenzyloxycarbonyl chloride,4-nitrobenzyloxycarbonyl chloride,chloroformic acid p-nitrobenzyl ester
78205
(4-nitrophenyl)methyl carbonochloridate
C1=CC(=CC=C1COC(=O)Cl)[N+](=O)[O-]
Infrared Spectrum Authentic
Refractive Index 1.5520 to 1.5560 (20°C, 589nm)
Quantity 25g
Color Yellow
CAS Min % 96.0
CAS Max % 100.0
Beilstein 06, 452
Formula Weight 215.59
Physical Form Liquid After Melting
Percent Purity ≥96.0%
Chemical Name or Material 4-Nitrobenzyl chloroformate, 97%
Show More Show Less

MOISTURE SENSITIVE

Danger