Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrobenzylamine hydrochloride, 94%

Catalog No. p-7025988
Click to view available options
Quantity:
1 g
5 g
25 g

CAS: 18600-42-5 | C7H9ClN2O2 | 188.611 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18600-42-5
Molecular Formula C7H9ClN2O2
Molecular Weight (g/mol) 188.611
MDL Number MFCD00012863
InChI Key SMIXZZMSWYOQPW-UHFFFAOYSA-N
Synonym 4-nitrobenzylamine hydrochloride, 4-nitrophenyl methanamine hydrochloride, p-nitrobenzylamine hydrochloride, 4-nitrobenzylamine hcl, pubchem12455, acmc-209enh, 4-nitrobenzylamine hcl salt, ksc493k2b, p-nitrobenzylaminehydrochloride, 4-nitrobenzylaminehydrochloride
PubChem CID 11252467
IUPAC Name (4-nitrophenyl)methanamine;hydrochloride
SMILES C1=CC(=CC=C1CN)[N+](=O)[O-].Cl

Specifications

Melting Point 255°C to 257°C (decomposition)
Quantity 1 g
Beilstein 3629994
Formula Weight 188.62
Percent Purity 97%
Chemical Name or Material 4-Nitrobenzylamine hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 242-441-3
RTECSNumber DP6705000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.