Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrocatechol 98.0+%, TCI America™
SDP

Catalog No. N027325G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 3316-09-4
Molecular Formula C6H5NO4
Molecular Weight (g/mol) 155.11
MDL Number MFCD00007242
InChI Key XJNPNXSISMKQEX-UHFFFAOYSA-N
Synonym 4-nitrocatechol, 4-nitropyrocatechol, 1,2-dihydroxy-4-nitrobenzene, 4-nitro-1,2-benzenediol, 3,4-dihydroxy-1-nitrobenzene, unii-sw60ng75en, sw60ng75en, 1,2-benzenediol, 4-nitro, nitrocatechol mix of isomers, 4nc
PubChem CID 3505109
ChEBI CHEBI:16318
IUPAC Name 4-nitrobenzene-1,2-diol
SMILES C1=CC(=C(C=C1[N+](=O)[O-])O)O

Specifications

Melting Point 175°C
Quantity 25 g
Formula Weight 155.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrocatechol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.