Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitrochalcone, 99%, Thermo Scientific™

Catalog No. p-173437
Click to view available options
Quantity:
25 g
Packaging:
Glass Bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1222-98-6
Molecular Formula C15H11NO3
Molecular Weight (g/mol) 253.257
MDL Number MFCD00007382
InChI Key WDZGGAFMGIOIQS-DHZHZOJOSA-N
Synonym 4-nitrochalcone, p-nitrostyryl phenyl ketone, chalcone, 4-nitro, p-nitrobenzylideneacetophenone, trans-4-nitrochalcone, 4-nitrocalone italian, unii-yt67d88ody, 3-4-nitrophenyl-1-phenyl-2-propen-1-one, p-nitrobenzylidene acetophenone, ccris 1669
PubChem CID 5377323
IUPAC Name (E)-3-(4-nitrophenyl)-1-phenylprop-2-en-1-one
SMILES C1=CC=C(C=C1)C(=O)C=CC2=CC=C(C=C2)[N+](=O)[O-]

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 158°C to 164°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula O2NC6H4CH=CHCOC6H5
Quantity 25 g
Beilstein 07, 482
Formula Weight 253.26
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 4-Nitrochalcone, 99%
Show More Show Less
EINECSNumber 214-949-5
RTECSNumber FL7075500
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.