Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 4-Nitrocinnamaldehyde, predominantly trans, 98%

Manufacturer:  Alfa Aesar™ A1146706

Quantity 5g

 View more versions of this product

Catalog No. AAA1146706


  Check Availability
Add to cart

Specifications

Description

Applications
Doebner-Miller reaction the 4- nitrocinnamaldehyde and 2-methylaniline in concentrated HC1 give the corresponding 8-methyl-2-phenylquinoline (3: R = 4 ft.-N02) directly. The asymmetric Friedel-Crafts-type alkylation in aqueous media reaction of 4-Nitrocinnamaldehydr with N-methyl indole using trifluoroacetic acid (TFA) salt of the PEG-PS-supported prolyl peptide having the polyleucine tether as a catalyst. 4-Nitrocinnamaldehyde has been used in the preparation of 2, 2 ft.-[(E)-3-(4-nitrophenyl) prop-2-ene-1,1-diyl] bis(3-hydroxy-5, 5-dimethylcyclohex-2-en-1-one).

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
Description & Specifications

Chemical Identifiers

1734-79-8
177.16
1565424
C9H7NO3
MFCD00007379
4-Nitrocinnamaldehyde, predominantly trans

Specifications

Characteristic
5g
C9H7NO3
MFCD00007379
4-Nitrocinnamaldehyde, predominantly trans
138° to 142°C
1734-79-8
177.16
1565424
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.

Warning

RTECSNumber : GD6650000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit