Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 4-Nitrofurantoin metabolite Monoclonal Antibody (1G7)
GREENER_CHOICE

Catalog No. PIMA549744
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549744 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549744 Supplier Invitrogen™ Supplier No. MA549744
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Antibody tested in indirect ELISA.

4-Nitrofurantoin is an antibiotic used to treat urinary tract infections. It works by stopping the growth of bacteria. When 4-Nitrofurantoin is ingested, it is metabolized in the liver and excreted in the urine. One of the metabolites produced is 4-Nitrophenol. This metabolite has been found to be genotoxic and mutagenic in some studies, and is therefore a concern for potential adverse effects on human health.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 4-Nitrofurantoin metabolite
Applications ELISA, Immunohistochemistry, Lateral Flow, SDS-Page
Classification Monoclonal
Clone 1G7
Concentration 1.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Host Species Mouse
Immunogen Conjugate of 4-CPAHD hapten and KLH.
Purification Method Protein G
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.