Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ 4-Nitrofurazone metabolite Monoclonal Antibody (6)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA549743
Description
Antibody tested in indirect ELISA.
4-Nitrofurazone is an antibiotic and antiprotozoal agent that is commonly used in veterinary medicine. It works by disrupting the bacterial cell wall and inhibiting the growth of protozoa. The metabolite of 4-Nitrofurazone is nifurtimox, which has antiprotozoal and antitumor properties. Nifurtimox has been used to treat Chagas disease, a parasitic infection that affects millions of people worldwide.
Specifications
4-Nitrofurazone metabolite | |
Monoclonal | |
6.8 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Conjugate of 4-CPSEM hapten and KLH. | |
200 μg | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry, Lateral Flow, SDS-Page | |
6 | |
Unconjugated | |
Mouse | |
Protein G | |
RUO | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction