Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Nitroindole 98%, ACROS Organics™

Click to view available options
Quantity:
2.5g
Packaging:
Glass bottle
4769-97-5
C8H6N2O2
162.15
MFCD00010056
LAVZKLJDKGRZJG-UHFFFAOYSA-N
4-nitroindole,indole, 4-nitro,1h-indole, 4-nitro,pubchem7440,4-nitro-1 h-indole,#,maybridge1_006408,acmc-209ka5,ksc236o2l
145766
4-nitro-1H-indole
C1=CC2=C(C=CN2)C(=C1)[N+](=O)[O-]
4769-97-5
C8H6N2O2
162.15
MFCD00010056
LAVZKLJDKGRZJG-UHFFFAOYSA-N
4-nitroindole,indole, 4-nitro,1h-indole, 4-nitro,pubchem7440,4-nitro-1 h-indole,#,maybridge1_006408,acmc-209ka5,ksc236o2l
145766
4-nitro-1H-indole
C1=CC2=C(C=CN2)C(=C1)[N+](=O)[O-]
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 203°C to 207°C
Quantity 2.5g
Color Yellow
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Solubility Information Solubility in water: insoluble
Formula Weight 162.15
Physical Form Granular Powder
Percent Purity ≥97.5%
Chemical Name or Material 4-Nitroindole, 98%
Show More Show Less

GHS H Statement:
Suspected of causing genetic defects.

GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.